This product is now discontinued, View Related Products
Low-Cost Specific Gravity Meters
Low-cost meter are ideal for general-purpose testing
Meters are simple to use—just dip probe into test liquid, then rinse with clean water after each use. Sample contamination due to inadequate probe cleaning between measurements is virtually eliminated. Meters are made from ABS plastic and are factory calibrated to ensure accuracy. Custom ranges other than the ranges offered are available—call our Application Specialists for details.
$3,195.05 - $3,440.37CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Specific Gravity Min Specific Gravity Temperature Resolution
Availability Pricing
Explorer Technology Group DSG-11-075100-C-B Low-Cost Specific Gravity Meter, 0.750 to 1.000 SGU, °C model RK-25755-00 Mfr # DSG-11-075100-C-B
COMPARE
1 0.75 0.1C
$3,351.23CAD / Each
Explorer Technology Group DSG-11-100130-C-B Low-Cost Specific Gravity Meter, 1.000 to 1.300 SGU, °C model RK-25755-04 Mfr # DSG-11-100130-C-B
COMPARE
1.3 1 0.1C
$3,440.37CAD / Each
Explorer Technology Group DSG-11-130160-C-B Low-Cost Specific Gravity Meter, 1.300 to 1.600 SGU, °C model RK-25755-10 Mfr # DSG-11-130160-C-B
COMPARE
1.6 1.3 0.1C
$3,440.37CAD / Each
Explorer Technology Group DSG-11-075100-F-B Low-Cost Specific Gravity Meter, 0.750 to 1.000 SGU, °F model RK-25755-14 Mfr # DSG-11-075100-F-B
COMPARE
1 0.75 0.1F
$3,195.05CAD / Each
Explorer Technology Group DSG-11-100130-F-B Low-Cost Specific Gravity Meter, 1.000 to 1.300 SGU, °F model RK-25755-20 Mfr # DSG-11-100130-F-B
COMPARE
1.3 1 0.1F
$3,195.05CAD / Each