This product is now discontinued, View Related Products
Lock Out/Tag Out; Danger - Do Not Start
Tags comply with tagout requirements of the OSHA Lockout/Tagout standard 29 CFR 1910.147. Use tags to mark and identify the lockout situation to prevent the accidental or unexpected start up or release of stored energy when working on equipment and machinery. Tags feature top gabled corners and 3/8” (9.5-mm) hole. All tags are sold in packages of 25.
Compliance: OSHA 29 CFR 1910.147
$30.94 - $117.87CAD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material
Availability Pricing
Accuform MLT403CTP Tag, Danger Do Not Start, 5 7/8" X 3 1/8" Pf-cardstock, 25/pk RK-40503-21 Mfr # MLT403CTP
COMPARE
Polyolefin
$30.94CAD / Pkg of 25
Accuform MLT403PTP Tag, Danger Do Not Start, 5 7/8" X 3 3/8", Rv-plastic, 25/pk RK-40503-22 Mfr # MLT403PTP
COMPARE
Plastic
$75.14CAD / Pkg of 25
Accuform MLT403LTP Tag, Danger Do Not Start, 5 3/4" X 3 1/16", Hs-laminate, 25/pk RK-40503-23 Mfr # MLT403LTP
COMPARE
Laminate
$117.87CAD / Pkg of 25