Light-Duty Mixer
Light-Duty Mixer is engineered for mixing smaller batches of light to medium-light viscosity fluids at high speeds.
Light-duty laboratory mixers include:
3/8" dia x 12"L, 316 SS shaft
2-1/2" dia, 316 SS propeller
collet
10"L x 9/16" dia aluminum mounting rod
6-ft, three-wire cord with plug for 115 VAC models; for 220 VAC models, specify country of destination for correct line cord
$1,337.84CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
Constant-torque, light-duty mixer, 115 VAC RK-50402-00 Mfr # 6000
COMPARE
6000 16.8 115
$1,337.84CAD / Each