Lascar Electronics Compact USB Data Loggers, Carbon Monoxide
Direct-to-USB connection and easy-to-use functionality
  • User-programmable alarm thresholds
  • All data loggers come with software
Simply insert any of these compact, 4.5" (11.4 cm) L loggers into your computer USB port, name it, choose your required sampling rate, select high and low alarms if required, and set the logger start time. Once configured, the logger can be removed from the computer to log independently. Data is downloaded back onto the PC where it can be analyzed and viewed as a graph using included software.
Use model 23039-58 for a 0 to 1000 ppm range and model 23039-68 for a 0 to 300 ppm measurement range; both store more than 32,000 readings. An bright red LED will flash and an audible alarm will sound when the user-determined level is exceeded.
$238.32 - $243.11CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Lascar Electronics EL-USB-CO USB Carbon Monoxide Data Logger, 32,000+ readings, 3 month battery life RK-23039-58 Mfr # EL-USB-CO
COMPARE
$238.32CAD / Each
Lascar Electronics EL-USB-CO300 Carbon monoxide Data Logger, 0 to 300 ppm, 32,000+ readings, 3 month battery life RK-23039-68 Mfr # EL-USB-CO300
COMPARE
$243.11CAD / Each