This product is now discontinued, View Related Products
LaMotte 2020 Handheld Turbidity Meters
Redesigned for higher accuracy and wider range
  • Advanced microprocessor offers high-accuracy readings
  • Available with either EPA- or ISO-compliant optical systems
  • Battery operated for field and on-site testing convenience
Test municipal waters, food and beverage waters, or any aqueous solution where fluid clarity is important. These compact portable meters are equipped with a unique optics configuration to read low turbidity levels down to 0.05 NTU and high levels up to 4000 NTU. Large, menu-driven LCD features five measuring units, seven user-selectable languages, datalogging capabilities to 500 points, and USB interface for downloading data to your computer.
$1,601.57 - $1,617.05CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Turbidity Min Turbidity Turbidity Resolution
Availability Pricing
LaMotte 1970-EPA 2020  Handheld Turbidity Meter, EPA compliant RK-05563-22 Mfr # 1970-EPA
COMPARE
4000 0.01 0.01, 0.1, or 1 NTU
$1,601.57CAD / Each
LaMotte 1970-ISO 2020wi Handheld Turbidity Meter, ISO compliant RK-05563-24 Mfr # 1970-ISO
COMPARE
4000 0.01 0.01, 0.1, or 1 NTU
$1,617.05CAD / Each