Labconco CentriVap Concentrator Accessories
Optimize performance of your CentriVap concentrator
  • Suitable for use with all CentriVap benchtop and mobile system concentrators
CentriZap Strobe Light enables the user to shine a strobe light into the CentriVap's chamber to easily view samples while the rotor is spinning. Includes mounting holster and hardware for attachment to the right side of the CentriVap. CentriVap not included.
$1,127.14CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Condenser Type Evaporation Flask Size Max Speed (rpm)
Availability Pricing
Labconco Stobe Light for CentriVap Concentrator RK-28656-60 Mfr # 7464300
COMPARE
$1,127.14CAD / Each