This product is now discontinued, View Related Products
Lab Companion Plant Growth Chambers
Precise airflow and temperature monitoring for optimized plant growth
These programmable growth chambers provide precise temperature control to maximize your plant growth. The 10-step program allows you to set temperature, humidity, and illumination profile which can be repeated up to 999 times. Dual sensors monitor water level and a quick-response electronic sensor maintains required humidity levels. Efficient airflow design provides even and accurate heat dispersion. Overtemperature safety features protect plants from being exposed to excess heat by shutting off power. Open door alarm minimizes loss of heat and humidity. Inner glass doors allow you to view plants without impairing interior conditions.
Interior is constructed of 304 stainless steel (304 SS) for easy clean-up and resistance to dirt. Exterior is made of powder-coated stainless steel. Interior doors are made of tempered glass for added protection and safety. Each unit includes casters for easier movement during transport and installation.
$175.33 - $41,696.94CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (cu ft) Power (VAC) Description
Availability Pricing
Jeio Tech Lab Companion AAHA1011 Plant Growth Chamber, 300 L, 230 VAC/60 Hz RK-39353-00 Mfr # AAHA1011K
COMPARE
10.6 230 Plant Growth Chamber, 300 L, 230 VAC/60 Hz
$26,456.19CAD / Each
Jeio Tech Lab Companion AAHA1011 Plant Growth Chamber, 300 L, 230 VAC/50 Hz RK-39353-05 Mfr # AAHA1012K
COMPARE
10.6 230 Plant Growth Chamber, 300 L, 230 VAC/50 Hz
$24,008.89CAD / Each
Jeio Tech Lab Companion AAHA1011 Plant Growth Chamber with Humidity Control, 300 L, 230VAC, 60 Hz RK-39353-10 Mfr # AAHA1021K
COMPARE
10.6 230 Plant Growth Chamber with Humidity Control, 300 L, 230VAC, 60 Hz
$35,268.54CAD / Each
Jeio Tech Lab Companion AAHA1011 Plant Growth Chamber with Humidity Control, 300 L, 230VAC, 50 Hz RK-39353-15 Mfr # AAHA1022K
COMPARE
10.6 230 Plant Growth Chamber with Humidity Control, 300 L, 230VAC, 50 Hz
$31,224.08CAD / Each
Jeio Tech Lab Companion AAHA1011 Plant Growth Chamber with Humidity Control,1000 L, 230VAC, 60 Hz RK-39353-20 Mfr # AAHA1031K
COMPARE
35.3 230 Plant Growth Chamber with Humidity Control,1000 L, 230VAC, 60 Hz
$41,696.94CAD / Each
Jeio Tech Lab Companion AAHA1011 Plant Growth Chamber with Humidity Control,1000 L, 230VAC, 50 Hz RK-39353-25 Mfr # AAHA1032K
COMPARE
35.3 230 Plant Growth Chamber with Humidity Control,1000 L, 230VAC, 50 Hz
$41,696.94CAD / Each
Lab Companion AAAA1511 Perforated Shelf For 300-liter Plant Growth Chambers RK-39353-70 Mfr # AAAA1511
COMPARE
Perforated Shelf For 300-liter Plant Growth Chambers
$327.09CAD / Each
Lab Companion AAAA1512 Perforated Shelf For 1000-liter Plant Growth Chambers RK-39353-72 Mfr # AAAA1512
COMPARE
Perforated Shelf For 1000-liter Plant Growth Chambers
$530.42CAD / Each
Lab Companion EDA8220 Wire Shelf For 300-liter Plant Growth Chamber RK-39353-80 Mfr # EDA8220
COMPARE
Wire Shelf For 300-liter Plant Growth Chamber
$175.33CAD / Each
Lab Companion EDA8222 Wire Shelf For 1000-liter Plant Growth Chamber RK-39353-82 Mfr # EDA8222
COMPARE
Wire Shelf For 1000-liter Plant Growth Chamber
$332.99CAD / Each
Lab Companion AAAA1501 Plant Growth Chamber Recorder, 6-point, humidity sensor RK-39353-90 Mfr # AAAA1501
COMPARE
Plant Growth Chamber Recorder, 6-point, humidity sensor
$8,177.31CAD / Each
Lab Companion AAAA1521 CoControl System For Plant Growth Chambers RK-39353-95 Mfr # AAAA1521
COMPARE
CoControl System For Plant Growth Chambers
$6,114.57CAD / Each
Showing 1- 12 of 12