Lab Companion Orbital Shakers
User-friendly control panel is easy to read and use
  • Platform stops and starts at center point every time, making repetitive tasks easy
  • Smooth acceleration and deceleration helps prevent spills from flasks or tubes
  • Low-profile design minimizes vibration and sliding due to high speed shaking
  • Automatic speed adjustment keeps unit stable in case of unbalanced load, uneven surface, or external shock
  • Platform and accessories must be purchased separately
Orbital shakers have microprocessor PID control enabling reproducible results every time. The control panel is easily read with a bright clear screen and the durable membrane touch switch and knob make setting and adjusting the speed simple and easy. Speed can be set and displayed in increments of 1, 5, or 10 depending on user preference.
Maintenance-free motor and powerful drive system generates smooth and quiet shaking motion over a wide speed range and under heavy workloads. Built-in protection system from over-current, stalled platform, and audible/visible alarms for shaking speed deviation keep the unit worry free. RS-232 and USB ports provide option for external control and data collection.
Platform and accessories must be purchased separately
$6,253.07 - $9,214.58CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Min Speed (rpm) Power (VAC)
Availability Pricing
Jeio Tech Lab Companion OS 4000 Orbital Shaker, 20 to 500 rpm, 17.7" W x 17.1" D Platform; 120 V RK-86578-40 Mfr # AAH3D1365U
COMPARE
500 20 120
$6,253.07CAD / Each
Jeio Tech Lab Companion OS 4000 Orbital Shaker, 20 to 500 rpm, 17.7" W x 17.1" D Platform; 230 V RK-86578-41 Mfr # AAH3D1315K
COMPARE
500 20 230
$6,328.21CAD / Each
Jeio Tech Lab Companion OS 7100 Orbital Shaker, 30 to 500 rpm, 29.6" W x 20.5" D Platform; 120 V RK-86578-42 Mfr # AAH3D1465U
COMPARE
500 30 120
$8,659.11CAD / Each
Jeio Tech Lab Companion OS 7100 Orbital Shaker, 30 to 500 rpm, 29.6" W x 20.5" D Platform; 230 V RK-86578-43 Mfr # AAH3D1415K
COMPARE
500 30 230
$9,207.21CAD / Each
Jeio Tech Lab Companion OS 7102 Orbital Shaker, 30 to 300 rpm, 29.6" W x 20.5" D Platform; 120 V RK-86578-44 Mfr # AAH3D1565U
COMPARE
300 30 120
$9,214.58CAD / Each