This product is now discontinued, View Related Products
Lab Companion Benchtop Incubating Shaker Kit
Shaking incubators and platforms bundled together for convenience and to save you money!
  • Worry-free operation—automatic speed adjustment keeps unit stable in case of unbalanced load, uneven surface, or external shock
  • Use the gentle orbital shaking motion from 10 to 500 rpm
  • Platform starts and stops at center position every time, making repetitive tasks easy to perform
  • Transparent lid with gas spring supports provides excellent visibility and easy sample access
  • RS-232 and USB ports for external control and data collection
Incubating shakers have an intuitive touch-screen LCD controller with easy-to-use icons and logical menus. Controls include automatic run with set point memory after a power interruption, auto-tuning, three-point temperature calibration, and a three-step adjustable fan for uniform temperature distribution and recovery after door opening. Safety features include audible/visible alarm activated by shaking speed deviation, protection system against over current or stalled platform, and dual mechanical overtemperature protection devices.
Maintenance-free motor generates quiet, uniform and powerful shaking motion for heavy workloads over a wide speed range. Smooth start and stop helps prevents spills from flasks or test tubes. Corrosion-resistant stainless steel interior is durable and easy to clean while the airtight silicone door seal provides excellent temperature uniformity.
$11,623.57 - $13,054.24CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Jeio Tech Lab Companion AAH23465U Complete Incubating Shaker Kit, 53 L, Universal; 120 VAC RK-25000-30 Mfr # AAH23465U
COMPARE
Complete Incubating Shaker Kit, 53 L, Universal; 120 VAC
$11,623.57CAD / Each
Jeio Tech Lab Companion AAH23665U Complete Incubating Shaker Kit, 83 L, Universal; 120 VAC RK-25000-34 Mfr # AAH23665U
COMPARE
Complete Incubating Shaker Kit, 83 L, Universal; 120 VAC
$13,054.24CAD / Each