This product is now discontinued, View Related Products
Koehler Manual Vacuum Distillation System
Equipped with digital temperature and vacuum displays for improved measurement reading
  • Conforms to ASTM D1160 and related specifications
Test the range of boiling points for petroleum and biofuel products with this vacuum distillation system. The sample is distilled at a reduced pressure and the initial and final boiling points are measured. A distillation curve can be generated relating volume percent distilled and the atmospheric equivalent boiling point temperature.
Main body of the system is constructed of an aluminum frame and cold-rolled steel walls with a clear protective door for added safety. Digital controller displays both overhead and flask temperature, along with vacuum reading. Built-in fan reduces cooling time of the distilling flask for shorter turnaround times between tests.
Optional Vacuum Pump Kit (59876-82) includes a vacuum pump, hose nozzle, hinged clamp, outlet filter, filter clamp, one liter of vacuum oil, connection tubing, and two hose clamps.
$30,777.64 - $31,175.46CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Koehler K80300 Manual Vacuum Distillation System, 115 VAC, 60 Hz RK-59876-80 Mfr # K80300
COMPARE
115
$31,175.46CAD / Each
Koehler K80390 Manual Vacuum Distillation System, 220-240 VAC, 50/60 Hz RK-59876-81 Mfr # K80390
COMPARE
220 / 240
$30,777.64CAD / Each