This product is now discontinued, View Related Products
Koehler Fluorescent Indicator Adsorption Apparatus
Quick column connection for fast setup and analysis
  • Conforms to ASTM D1319 specifications
  • Conduct up to six FIA analyses simultaneously
Determine the total volume percent of aromatic, olefins, and saturates in petroleum fractions with this fluorescent indicator adsorption apparatus. Multiposition air manifold features independently operated gauges, pressure regulators, and ball O-ring joints for individual pressure control at each column. An integrated electric vibration system facilitates the dry gel packing of the columns.
After the sample is introduced and adsorbed into the column, fluorescent dye indicators mark the boundaries of aromatics, olefins, and saturates, which are visible with a UV lamp. Calculate the volume percentage using the length of each zone in the column.
$22,999.62CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Koehler K41506 Fluorescent Indicator Absorption Apparatus, 115 VAC, 50/60 Hz RK-59876-85 Mfr # K41506
COMPARE
115
$22,999.62CAD / Each
Koehler K41596 Fluorescent Indicator Absorption Apparatus, 230 VAC, 50/60 Hz RK-59876-86 Mfr # K41596
COMPARE
230
$22,999.62CAD / Each