This product is now discontinued, View Related Products
Koehler Cloud and Pour Point Refrigerated Baths
Cascade refrigeration system provides reliable, long-term service
  • Conforms to ASTM D2500 and related specifications
Determine the temperature when the smallest observable cluster of crystals occurs upon cooling, in accordance with ASTM D2500. Benchtop unit features three jacketed mechanically refrigerated baths, each preset to a different temperature: -30, 0, and 30°F (-35, -18, and -1°C). Each bath has a phenolic top plate with ports for a thermometer and four copper test jackets. Bath interior is constructed of stainless steel; cabinet is constructed of polyester-epoxy finished steel.
$25,939.03 - $34,580.46CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
Koehler K46100 Cloud and Pour Point Bath, 115 VAC RK-59861-10 Mfr # K46100
COMPARE
115 Cloud and Pour Point Bath, 115 VAC
$34,580.46CAD / Each
Koehler K46100 Cloud and Pour Point Bath, 220-240 VAC RK-59861-15 Mfr # K46195
COMPARE
220 / 240 Cloud and Pour Point Bath, 220-240 VAC
$25,939.03CAD / Each