This product is now discontinued, View Related Products
Cole-Parmer® TELOS® SPE-Format NH2 Flash Chromatography Columns
Ideal for first stage clean-up columns post synthesis using a vacuum manifold
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
Easily connect to flash chromatography system using the Luer-slip connection. Use columns on a rack or stand using gravity flow or with vacuum processing stations. All columns are made from polypropylene and come with polyethylene frits.
$178.28 - $851.62CAD / Pkg of 20
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (g)
Availability Pricing
COMPARE
2
$178.28CAD / Pkg of 20
COMPARE
25 5
$306.47CAD / Pkg of 20
COMPARE
70 10
$446.44CAD / Pkg of 16
COMPARE
70 20
$668.92CAD / Pkg of 16
COMPARE
25
$732.27CAD / Pkg of 16
COMPARE
150 50
$779.42CAD / Pkg of 8
COMPARE
150 70
$851.62CAD / Pkg of 8
Showing 1- 7 of 7