Cole-Parmer® TELOS® Silica Flash Chromatography Columns with Luer-lok® inlet and Luer-slip outlet
Use with systems from ISCO® and Buchi®
  • Optimized column geometry provides maximum recovery
  • Excellent for purification of a wide range of synthetic and natural compounds
Make connections to flash chromatography instruments quick and easy with Luer connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.
$128.18 - $266.68CAD / Pkg of 20
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Bed Mass (g)
Availability Pricing
COMPARE
4
$198.91CAD / Pkg of 20
COMPARE
12
$212.17CAD / Pkg of 20
COMPARE
20
$266.68CAD / Pkg of 20
COMPARE
40
$153.23CAD / Pkg of 10
COMPARE
45
$210.69CAD / Pkg of 10
COMPARE
80
$128.18CAD / Pkg of 5
COMPARE
120
$143.66CAD / Pkg of 5
COMPARE
220
$137.03CAD / Pkg of 1
COMPARE
330
$141.45CAD / Pkg of 1
Showing 1- 9 of 9