Cole-Parmer® TELOS® neo™ SPE Fixed 96-Well Plates
Modular design lets you simply remove any unwanted wells and add sealing strips
  • Quickly obtain clean extracts and reduce time spent on method development
  • Faster method development saves you money
  • Water-wettable polymer is not affected by drying out
Cole-Parmer® TELOS® neo™ populated plates come with 96 wells already in place—ideal for high throughput assays. Polymer-based sorbents have no silanol interactions, excellent flow characteristics, and pH stability providing highly reproducible results. Elution volumes are kept to a minimum by allowing maximum interaction between the analyte and sorbent.
$402.24 - $449.38CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sorbent Applications Bed Mass (mg)
Availability Pricing
COMPARE
WCX Mixed-mode weak cation exchange 30
$449.38CAD / Each
COMPARE
WAX Mixed-mode weak anion exchange 30
$431.70CAD / Each
COMPARE
PAX Mixed-mode strong anion exchange 30
$431.70CAD / Each
COMPARE
WCX Mixed-mode weak cation exchange 10
$412.55CAD / Each
COMPARE
PCX Mixed-mode strong cation exchange 10
$412.55CAD / Each
COMPARE
PCX Mixed-mode strong cation exchange 30
$449.38CAD / Each
COMPARE
PAX Mixed-mode strong anion exchange 10
$431.70CAD / Each
COMPARE
PRP Nonpolar 10
$402.24CAD / Each
Showing 1- 8 of 8