Kinesis® Screw Caps for Screw Cap Vials
No more damaged autosampler needles or sample loss
  • Ensure septa is not deformed due to overtightening and extend the life of your needle
  • Prevent sample evaporation due to poor vial-septa-cap sealing
  • UltraClean™ high-purity septa are optimized for GC and headspace analysis
  • Select caps with UltraBond­® septum to avoid sample contamination
Select from polypropylene (PP), polyethylene (PE) or magnetic silver caps with various septum materials. Caps are available in different sizes, colors, septum materials, and open or closed configurations to suit your application. UltraBond cap and septum form a one-piece unit with a bonding process without adhesives for contamination-free results. Pre-slit septa prevent a vacuum forming inside the vial.
$26.37 - $1,410.03CAD / Pkg of 1000
50 variations of this product are available.
See All Product Options Below
50 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Diameter (mm) Cap Material Septum Type
Availability Pricing
COMPARE
13 Polypropylene Silicone white/PTFE red
$433.18CAD / Pkg of 1000
COMPARE
13 Magnetic silver Silicone/Red PTFE
$879.61CAD / Pkg of 1000
COMPARE
8 Polypropylene Natural rubber/TEF transparent
$338.88CAD / Pkg of 1000
COMPARE
8 Polypropylene Natural rubber/TEF red
$355.09CAD / Pkg of 1000
COMPARE
8 Polypropylene Silicone white/PTFE red
$462.64CAD / Pkg of 1000
COMPARE
8 Polypropylene None
$70.58CAD / Pkg of 1000
COMPARE
8 Polypropylene None
$76.62CAD / Pkg of 1000
COMPARE
8 Polypropylene None
$75.14CAD / Pkg of 1000
COMPARE
8 Polypropylene None
$92.82CAD / Pkg of 1000
COMPARE
10 Polypropylene Natural rubber/PTFE transparent
$405.18CAD / Pkg of 1000
COMPARE
10 Polypropylene None
$206.27CAD / Pkg of 1000
COMPARE
13 Polypropylene Butyl red/PTFE gray
$394.87CAD / Pkg of 1000
COMPARE
13 Polypropylene Silicone white/PTFE red
$717.54CAD / Pkg of 1000
COMPARE
13 Polypropylene Natural rubber/TEF transparent
$453.80CAD / Pkg of 1000
COMPARE
13 Polypropylene None
$143.66CAD / Pkg of 1000
COMPARE
13 Polypropylene None
$184.17CAD / Pkg of 1000
COMPARE
15 Polypropylene Butyl red/PTFE gray
$564.31CAD / Pkg of 1000
COMPARE
15 Polypropylene Natural rubber/TEF transparent
$495.06CAD / Pkg of 1000
COMPARE
15 Polypropylene Silicone white/PTFE red
$900.24CAD / Pkg of 1000
COMPARE
15 Polypropylene None
$26.37CAD / Pkg of 100
Showing 1 - 20 of 50