This product is now discontinued, View Related Products
Kinesis Screw Caps for EPA Vials
Caps come preassembled with silicone/PTFE septa permanently bonded to the cap to reduce the chance of contamination
  • Select from open (injection), closed (storage), or SURE-Clean™ caps, depending on your application
All caps come with silicone/PTFE bonded septa. Each septum is 3.2 mm thick and had hardness rating of 45° shore A.
$940.76 - $1,190.50CAD / Pkg of 1000
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Cap Type Septum Type
Availability Pricing
COMPARE
Screw cap Bonded silicone/PTFE bonded
$978.33CAD / Pkg of 1000
COMPARE
Screw cap Bonded silicone/PTFE bonded
$940.76CAD / Pkg of 1000
COMPARE
SURE-Clean™ screw cap Bonded silicone/PTFE bonded
$1,190.50CAD / Pkg of 1000