This product is now discontinued, View Related Products
Cole-Parmer®+ LCGC Certified Limited-Volume Snap Top Polypropylene Vials with Caps
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Snap-top vials come with snap caps that provide a secure seal to minimize evaporation, even with volatile samples. An audible click ensures a secure seal has been formed.
Limited volume vials are excellent for handling small-volume samples while delivering low residual volume (<8 µL for 300 µL vials; ,80 µL for 700 µL vials). Polypropylene (PP) is ideal for samples where glass is not appropriate; as with samples that adhere to glass.
$77.35 - $109.62CAD / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Septum Type Volume (mL)
Availability Pricing
COMPARE
LCGC Limited-Volume Snap Top Vials, Polypropylene, 11 mm Short Thread, 0.3 mL, PTFE/Silicone Septum; 100/pk PTFE/silicone 0.3
$77.35CAD / Pkg of 100
COMPARE
LCGC Limited-Volume Snap Top Vials, Polypropylene, 11 mm Short Thread, 0.3 mL, Pre-Slit PTFE/Silicone Septum; 100/pk Pre-slit PTFE/silicone 0.3
$109.62CAD / Pkg of 100
COMPARE
LCGC Limited-Volume Snap Top Vials, Polypropylene, 11 mm Short Thread, 0.7 mL, PTFE/Silicone Septum; 100/pk PTFE/silicone 0.7
$77.35CAD / Pkg of 100
COMPARE
LCGC Limited-Volume Snap Top Vials, Polypropylene, 11 mm Short Thread, 0.7 mL, Pre-Slit PTFE/Silicone Septum; 100/pk Pre-slit PTFE/silicone 0.7
$109.62CAD / Pkg of 100