This product is now discontinued, View Related Products
Heraeus Noblelight Hollow Cathode Lamps, Europium
Achieve reliable atomic absorption results with high-performance lamps for your spectrometers
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
Heraeus Noblelight europium (Eu) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp’s lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer®, Varian/Agilent, and Unicam systems.
$473.11 - $934.13CAD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
COMPARE
Europium Uncoded Hollow Cathode Lamp, europium, 37 mm, uncoded
$583.09CAD / Pkg of 1
COMPARE
Europium Self Reversal Hollow Cathode Lamp, europium, self reversal
$473.11CAD / Pkg of 1
COMPARE
Europium Unicam Hollow Cathode Lamp, europium, 37 mm, Unicam coded
$663.03CAD / Pkg of 1
COMPARE
Europium Varian Hollow Cathode Lamp, europium, 37 mm, Varian coded
$663.03CAD / Pkg of 1
COMPARE
Europium Perkin Elmer Standard Hollow Cathode Lamp, europium, 50 mm, Perkin Elmer Standard coded
$738.17CAD / Pkg of 1
COMPARE
Europium Perkin Elmer Lumina Hollow Cathode Lamp, europium, 50 mm, Perkin Elmer Lumina coded
$934.13CAD / Pkg of 1