Cole-Parmer® EPA Glass Vials
Meets EPA requirements for testing environmental contaminants
  • Made of Type I borosilicate glass for testing of potentially harmful environmental contaminants in water or soil samples
  • Vials meet the requirements of the US Environmental Protection Agency (EPA) for contamination-free containers
  • Choose from 20-, 40-, or 60-mL vials or vial kits for your specific application
These flat bottom EPA vials are free of substances that might influence sample analysis. Chemically inert type I borosilicate glass is ideal for soil and water samples. Vials are available in clear glass or amber glass for light sensitive samples. Select from standard EPA vials, vials with a certificate of cleanliness, preassembled vials with caps, or precleaned and preassembled vials with caps.
$618.82 - $1,989.08CAD / Pkg of 1000
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Description Tube Color Volume (mL)
Availability Pricing
COMPARE
EPA Screw Neck Vial, 30 mL, 24mm, Amber Glass; 1000/pk Amber 30
$1,709.13CAD / Pkg of 1000
COMPARE
EPA Screw Neck Vial, 30 mL, 24mm, Glass; 1000/pk Glass 30
$618.82CAD / Pkg of 1000
COMPARE
EPA Screw Neck Vial, 40 mL, 24mm, Glass, Certified; 1000/pk Glass 40
$876.67CAD / Pkg of 1000
COMPARE
EPA Screw Neck Vial, 60 mL, 24mm, Amber Glass; 1000/pk Amber 60
$1,989.08CAD / Pkg of 1000
COMPARE
EPA Screw Neck Vial, 60 mL, 24mm, Glass; 1000/pk Glass 60
$870.77CAD / Pkg of 1000