Kinesis® EPA Caps and Septa for 24 mm EPA Vials
Meets EPA requirements for testing environmental contaminants
  • Septa meet the requirements of the US Environmental Protection Agency (EPA)
  • Caps are constructed of durable white polypropylene
  • Select caps with UltraBond® septum to avoid sample contamination
These white polypropylene (PP) caps and EPA septa are free of substances that might influence sample analysis for testing of potentially harmful environmental contaminants in water or soil samples. Select from septa only, cap only, or cap and septa combination.
$375.71 - $1,362.89CAD / Pkg of 1000
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description Septum Type Special Features
Availability Pricing
COMPARE
EPA Screw Cap, 24mm, White Polypropylene, Open, Silicone White/PTFE Beige Septa; 1000/pk Silicone White/PTFE Beige Septa Open Cap
$1,362.89CAD / Pkg of 1000
COMPARE
EPA Screw Cap, 24mm, White Polypropylene, Closed, Silicone White/PTFE Beige Septa; 1000/pk Silicone White/PTFE Beige Septa Closed Cap
$1,349.63CAD / Pkg of 1000
COMPARE
EPA Screw Cap, 24mm, Polypropylene, Closed, UltraBond Silicone White/PTFE Beige Septa; 1000/pk Ultrabond Silicone White/PTFE Beige Septa UltraBond
$1,228.81CAD / Pkg of 1000
COMPARE
EPA Screw Cap, 24mm, Polypropylene, Open, UltraBond Silicone White/PTFE Beige Septa; 1000/pk Ultrabond Silicone White/PTFE Beige Septa UltraBond
$1,233.23CAD / Pkg of 1000
COMPARE
EPA Septa, 24mm, Silicone Natural/PTFE Beige; 1000/pk Silicone Natural/PTFE Beige Septa only
$375.71CAD / Pkg of 1000
COMPARE
EPA Screw Cap, 24mm, White Polypropylene, Open, Butyl Red/PTFE Grey Septa; 1000/pk Butyl Red/PTFE Grey Open Cap
$858.99CAD / Pkg of 1000
COMPARE
EPA Screw Cap, 24mm, White Polypropylene, Closed, Butyl Red/PTFE Grey Septa; 1000/pk Butyl Red/PTFE Grey Closed Cap
$693.97CAD / Pkg of 1000
Showing 1- 7 of 7