Kinesis® Disposable Syringes, Polypropylene
Avoid sample contamination
  • Free of latex, plasticizers, PVC, and rubber or synthetic gaskets
  • Reduce contamination inside the barrel with silicone and oil lubricant free construction
The two-part, all-plastic syringes are compatible with a wide range of chemicals. The barrel and plunger are made of inert, chemical-resistant polypropylene (PP). Transparent barrels and permanent graduations simplify the monitoring and inspection of liquids. Syringes are nonsterile and have a standard conical fitting with 6% (Luer) taper.
These disposable syringes are available with two different connector types: Luer-Lok® or Luer-slip. Luer-Lok syringes have a twist-and-lock mechanism to create a secure fitting connection, making them a safe choice for high back pressure liquids in filter applications. Luer-slip syringes slide into place and are held by friction—ideal for low-pressure applications.
Each manufactured batch is HPLC controlled and supplied with a proper certificate upon request.
$27.41 - $68.37CAD / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Syringe Capacity (mL) Syringe Type
Availability Pricing
COMPARE
5 Luer-Lok®
$68.37CAD / Pkg of 100
COMPARE
20 Luer slip
$62.47CAD / Pkg of 100
COMPARE
2 Luer slip
$27.41CAD / Pkg of 100
COMPARE
5 Luer slip
$34.33CAD / Pkg of 100