This product is now discontinued, View Related Products
Kimcote Safety-Coated, Heavy-Duty Solution Bottle
Controls spills in the event of glassware breakage
  • Ideal for chemical storage
  • Autoclavable to120°C (250°F)
Ultra-clear safety-coated glassware helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware is ideal for chemical storage, transportation, and handling of hazardous chemicals, harmful biologicals, and acids. Glassware has a unique nonslip surface when wet or dry. Durable bottles are tooled to fit a #12 size stopper. Five gallon size is designed from Federal Spec DD-B-597. Autoclavable to120°C (250°F).
$845.73 - $1,535.27CAD / Case of 1
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (L) Material
Availability Pricing
DWK Life Sciences (Kimble) KC14950-25 Heavy-Duty Solution Bottle (Carboy) 9.5 L / 2.5 gal. RK-34523-29 Mfr # KC14950-25
COMPARE
9.5 Glass
$1,208.18CAD / Case of 1
DWK Life Sciences (Kimble) KC14950-35 Heavy-Duty Solution Bottle (Carboy) 13.2 L / 3.5 gal. RK-34523-31 Mfr # KC14950-35
COMPARE
13.2 Glass
$845.73CAD / Case of 1
DWK Life Sciences (Kimble) KC14950-500 Heavy-Duty Solution Bottle (Carboy) 19.0 L / 5.0 gal. RK-34523-33 Mfr # KC14950-500
COMPARE
19 Glass
$1,044.63CAD / Case of 1
DWK Life Sciences (Kimble) KC14950-120 Heavy-Duty Solution Bottle (Carboy) 45.5 L / 12.0 gal. RK-34523-35 Mfr # KC14950-120
COMPARE
45.5 Glass
$1,535.27CAD / Case of 0