KIMAX® Class A Colorware Glass Volumetric Flasks
Color coding provides quick and easy identification for specific applications
  • Constructed of low extractable borosilicate glass
  • Volumetric flask features matte finish marking area for use with an ordinary pencil
Calibrated "to contain," these flasks are available with markings in four colors, the KIMAX® colorware volumetric flasks are brighter in appearance than traditional glassware with white markings. These flasks are also commonly used to help reduce cross-contamination and for easy identification of glassware between labs.
These Class A volumetric flasks have a single graduation ring blasted on the neck and include a ground glass standard taper stopper. Calibrated to meet ASTM E 288, Class A unserialized specifications.
$178.28 - $878.88CAD / Pkg of 6
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Tolerance (mL) Description
Availability Pricing
DWK Life Sciences (Kimble) Green Coded Class A Volumetric Flask, 25 mL, 6/PK RK-34562-10 Mfr # 28014E-25
COMPARE
25 ±0.03 Green Coded Volumetric Flask, 25 mL, 6/pk
$490.64CAD / Pkg of 6
DWK Life Sciences (Kimble) Green Coded Class A Volumetric Flask, 50 mL, 6/PK RK-34562-12 Mfr # 28014E-50
COMPARE
50 ±0.05 Green Coded Volumetric Flask, 50 mL, 6/pk
$552.52CAD / Pkg of 6
DWK Life Sciences (Kimble) Green Coded Class A Volumetric Flask, 100 mL, 6/PK RK-34562-14 Mfr # 2.8014E-96
COMPARE
100 ±0.08 Green Coded Volumetric Flask, 100 mL, 6/pk
$552.52CAD / Pkg of 6
DWK Life Sciences (Kimble) Green Coded Class A Volumetric Flask, 250 mL, 6/PK RK-34562-16 Mfr # 2.8014E-246
COMPARE
250 ±0.12 Green Coded Volumetric Flask, 250 mL, 6/pk
$682.92CAD / Pkg of 6
DWK Life Sciences (Kimble) Green Coded Class A Volumetric Flask, 500 mL, 6/PK RK-34562-18 Mfr # 28014E-500
COMPARE
500 ±0.20 Green Coded Volumetric Flask, 500 mL, 6/pk
$854.57CAD / Pkg of 6
DWK Life Sciences (Kimble) Green Coded Class A Volumetric Flask, 1000 mL, 1/PK RK-34562-20 Mfr # 28014E-1000
COMPARE
32 ±0.30 Green Coded Volumetric Flask, 1000 mL, 1/pk
$178.28CAD / Pkg of 1
DWK Life Sciences (Kimble) Blue Coded Class A Volumetric Flask, 25 mL, 6/PK RK-34562-30 Mfr # 28014B-25
COMPARE
25 ±0.03 Blue Coded Volumetric Flask, 25 mL, 6/pk
$506.11CAD / Pkg of 6
DWK Life Sciences (Kimble) Blue Coded Class A Volumetric Flask, 50 mL, 6/PK RK-34562-32 Mfr # 28014B-50
COMPARE
50 ±0.05 Blue Coded Volumetric Flask, 50 mL, 6/pk
$571.68CAD / Pkg of 6
DWK Life Sciences (Kimble) Blue Coded Class A Volumetric Flask, 100 mL, 6/PK RK-34562-34 Mfr # 28014B-100
COMPARE
100 ±0.08 Blue Coded Volumetric Flask, 100 mL, 6/pk
$571.68CAD / Pkg of 6
DWK Life Sciences (Kimble) Blue Coded Class A Volumetric Flask, 250 mL, 6/PK RK-34562-36 Mfr # 28014B-250
COMPARE
250 ±0.12 Blue Coded Volumetric Flask, 250 mL, 6/pk
$707.23CAD / Pkg of 6
DWK Life Sciences (Kimble) Blue Coded Class A Volumetric Flask, 500 mL, 6/PK RK-34562-38 Mfr # 28014B-500
COMPARE
500 ±0.20 Blue Coded Volumetric Flask, 500 mL, 6/pk
$878.88CAD / Pkg of 6
DWK Life Sciences (Kimble) Blue Coded Class A Volumetric Flask, 1000 mL, 1/PK RK-34562-40 Mfr # 28014B-1000
COMPARE
1000 ±0.30 Blue Coded Volumetric Flask, 1000 mL, 1/pk
$183.44CAD / Pkg of 1
DWK Life Sciences (Kimble) Red Coded Class A Volumetric Flask, 100 mL, 6/PK RK-34562-54 Mfr # 28014R-100
COMPARE
100 ±0.08 Red Coded Volumetric Flask, 100 mL, 6/pk
$571.68CAD / Pkg of 6
DWK Life Sciences (Kimble) Red Coded Class A Volumetric Flask, 500 mL, 6/PK RK-34562-58 Mfr # 28014R-500
COMPARE
500 ±0.20 Red Coded Volumetric Flask, 500 mL, 6/pk
$878.88CAD / Pkg of 6
DWK Life Sciences (Kimble) Red Coded Class A Volumetric Flask, 1000 mL, 1/PK RK-34562-60 Mfr # 28014R-1000
COMPARE
1000 ±0.30 Red Coded Volumetric Flask, 1000 mL, 1/pk
$183.44CAD / Pkg of 1
DWK Life Sciences (Kimble) Yellow Coded Class A Volumetric Flask, 100 mL, 6/PK RK-34562-74 Mfr # 28014Y-100
COMPARE
100 ±0.08 Yellow Coded Volumetric Flask, 100 mL, 6/pk
$571.68CAD / Pkg of 6
DWK Life Sciences (Kimble) Yellow Coded Class A Volumetric Flask, 500 mL, 6/PK RK-34562-78 Mfr # 28014Y-500
COMPARE
500 ±0.20 Yellow Coded Volumetric Flask, 500 mL, 6/pk
$878.88CAD / Pkg of 6
DWK Life Sciences (Kimble) Yellow Coded Class A Volumetric Flask, 1000 mL, 1/PK RK-34562-80 Mfr # 28014Y-1000
COMPARE
1000 ±0.30 Yellow Coded Volumetric Flask, 1000 mL, 1/pk
$183.44CAD / Pkg of 1
Showing 1- 18 of 18