Kanomax Climomaster 6501 Hot-Wire Anemometer Probes
Research-grade accuracy for airflow measurement—ideal for laboratory, IAQ, and critical environment monitoring
  • Smart probe technology lets you change probes without recalibrating the main unit—order meter separately to create your system
Unidirectional probes are economical and great for applications where the direction of the airflow is known. Omnidirectional probes can sense airflow from all directions—perfect for turbulent airflow applications.
$673.34 - $2,096.63CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kanomax Airflow Measurement Probe for Velocity, Temperature, and Humidity; RK-10007-17 Mfr # 6531-2G
COMPARE
Airflow Measurement Probe for Velocity, Temperature, and Humidity;
$1,093.26CAD / Each
Kanomax Airflow Measurement Probe for Velocity, Temperature, and Humidity; Omnidirectional RK-10007-18 Mfr # 6533-2G
COMPARE
Airflow Measurement Probe for Velocity, Temperature, and Humidity; Omnidirectional
$2,005.28CAD / Each
Kanomax Airflow Measurement Probe for Velocity and Temperature; Unidirectional RK-10007-19 Mfr # 6541-2G
COMPARE
Airflow Measurement Probe for Velocity and Temperature; Unidirectional
$673.34CAD / Each
Kanomax Airflow Measurement Probe for Velocity and Temperature; Omnidirectional, Needle RK-10007-27 Mfr # 6542-2G
COMPARE
Airflow Measurement Probe for Velocity and Temperature; Omnidirectional, Needle
$1,184.61CAD / Each
Kanomax Airflow Measurement Probe for Velocity and Temperature; Omnidirectional, Sphere RK-10007-28 Mfr # 6543-2G
COMPARE
Airflow Measurement Probe for Velocity and Temperature; Omnidirectional, Sphere
$1,458.66CAD / Each
Kanomax Airflow Measurement Probe for Velocity; Omnidirectional, Mini L Shape SKU 1000729 RK-10007-29 Mfr # 6551-2G
COMPARE
Airflow Measurement Probe for Velocity; Omnidirectional, Mini L Shape
$2,096.63CAD / Each
Kanomax Airflow Measurement Probe for Velocity; Omnidirectional, Mini L Shape SKU 1000736 RK-10007-36 Mfr # 6552-2G
COMPARE
Airflow Measurement Probe for Velocity; Omnidirectional, Mini L Shape
$1,934.56CAD / Each
Kanomax Airflow Measurement Probe for Velocity and Temperature; Unidirectional, High Velocity RK-10007-37 Mfr # 6561-2G
COMPARE
Airflow Measurement Probe for Velocity and Temperature; Unidirectional, High Velocity
$1,093.26CAD / Each
Showing 1- 8 of 8