This product is now discontinued, View Related Products
Kanomax Airflow Transducer Probes
Work with the Kanomax airflow transducers
  • Built-in temperature compensation sensor in the probe provides more accurate measurements
  • Smart probe technology lets you change probes without recalibrating the transducer
$676.58 - $1,934.56CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kanomax Airflow Transducer Probe, Uni, Rod, 3" L; 30 cm Cable RK-10006-60 Mfr # 0962-00
COMPARE
Airflow Transducer Probe, Uni, Rod, 3\" L; 30 cm Cable
$698.39CAD / Each
Kanomax Airflow Transducer Probe, Uni, Rod, 4.25" L; No Cable RK-10006-61 Mfr # 0963-00
COMPARE
Airflow Transducer Probe, Uni, Rod, 4.25\" L; No Cable
$676.58CAD / Each
Kanomax Airflow Transducer Probe, Omni, Spherical, Tipped, 4" L; No Cable RK-10006-62 Mfr # 0965-00
COMPARE
Airflow Transducer Probe, Omni, Spherical, Tipped, 4\" L; No Cable
$1,099.52CAD / Each
Kanomax Airflow Transducer Probe, Omni, Spherical, 4" L, 25 m/s max; No Cable RK-10006-63 Mfr # 0965-01
COMPARE
Airflow Transducer Probe, Omni, Spherical, 4\" L, 25 m/s max; No Cable
$1,268.22CAD / Each
Kanomax Airflow Transducer Probe, Omni, Mini Straight, 2.125" L, 100 cm Cable RK-10006-64 Mfr # 0965-03
COMPARE
Airflow Transducer Probe, Omni, Mini Straight, 2.125\" L, 100 cm Cable
$1,099.52CAD / Each
Kanomax Airflow Transducer Probe, Omni, Mini Bent, 2.125 "L, 100 cm Cable RK-10006-65 Mfr # 0965-04
COMPARE
Airflow Transducer Probe, Omni, Mini Bent, 2.125 \"L, 100 cm Cable
$1,099.52CAD / Each
Kanomax Airflow Transducer Probe, Omni/Mini Straight/Sep ATC/2.125" L/100 cm Cable RK-10006-66 Mfr # 0965-07
COMPARE
Airflow Transducer Probe, Omni/Mini Straight/Sep ATC/2.125\" L/100 cm Cable
$1,268.22CAD / Each
Kanomax Airflow Transducer Probe, Omni/Mini Bent/Sep ATC/2.125" L, 100 cm Cable RK-10006-67 Mfr # 0965-08
COMPARE
Airflow Transducer Probe, Omni/Mini Bent/Sep ATC/2.125\" L, 100 cm Cable
$1,268.22CAD / Each
Kanomax Airflow Transducer Probe, Omni, Spherical, 4" L, 50 m/s max; No Cable RK-10006-68 Mfr # 0965-09
COMPARE
Airflow Transducer Probe, Omni, Spherical, 4\" L, 50 m/s max; No Cable
$1,849.84CAD / Each
Kanomax Airflow Transducer Probe, Omni, Spherical, 16.125" L, 50 m/s max; No Cable RK-10006-69 Mfr # 0965-10
COMPARE
Airflow Transducer Probe, Omni, Spherical, 16.125\" L, 50 m/s max; No Cable
$1,934.56CAD / Each
Showing 1- 10 of 10