This product is now discontinued, View Related Products
Kanomax Air Flow Transducers
Analog output makes it easy to integrate with other equipment
  • Purchase probes separately and choose the ones that meet your application needs!
  • Smart probe technology lets you change probes without recalibrating the transducer
Airflow transducers are designed for fixed installation—great for production line quality control and monitoring local exhaust ventilation system. Transducers feature 0 to 5 VDC or 4 to 20 mA analog output so you can send your data to other instruments such as data loggers, alarms, or controllers. Choose from a unit with a display for on-site
$1,183.50 - $1,522.01CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kanomax Airflow Transducer with Display RK-10006-01 Mfr # 6332D
COMPARE
Airflow Transducer with Display
$1,522.01CAD / Each
Kanomax Airflow Transducer without Display RK-10006-02 Mfr # 6332
COMPARE
Airflow Transducer without Display
$1,183.50CAD / Each