Justrite Corrosive/Acid Plastic Safety Cabinets
All-polyethylene construction means there are no metal parts to corrode
  • Door accepts a padlock for security against unauthorized use
Store your highly aggressive liquids such as phenol, nitric acid, and sulfuric acid in these all high-density polyethylene (HDPE) cabinets. These cabinets feature hinges, door handle, and internal latching mechanism made of polyethylene There are no metal parts to corrode. Welded 2" (51 mm) sump at the bottom of the cabinet contains accidental spills.
Toe kick on undercounter model offers improved ergonomics—less leaning. Cabinets also includes an external 2" NPT vent/exhaust connection, leveling feet on all four corners, and reflective warning label in three languages. Door accepts optional padlock.
$12,825.86 - $15,283.47CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Height (in) Material
Availability Pricing
Justrite 24010 Corrosives/Acids Plastic Safety Cabinet, Undercounter, Manual Close, 2 Doors, 24 Gal RK-81790-51 Mfr # 24010
COMPARE
24 35 3/4 HDPE
$12,825.86CAD / Each
Justrite 24015 Corrosives/Acids Plastic Safety Cabinet, Stand Alone, Manual Close, 2 Doors, 24 Gal RK-81790-52 Mfr # 24015
COMPARE
24 35 3/4 HDPE
$15,283.47CAD / Each