Justrite Sure-Grip® EX Flammable Storage Safety Cabinets
Protect workers while reducing fire risks
  • Independently fire tested and approved by FM Approvals
  • OSHA and NFPA compliant
Cabinets feature 18-gauge steel, all-welded, double-wall, 1 1/2" insulating air barrier construction for protection and durability. Adjustable, patented SpillSlope® galvanized steel shelves direct spills to back and bottom of 2” (51mm) leakproof sump.
All models feature self-latching doors with U-loc™ padlockable handle and 3-point stainless steel bullet latching system to ensure complete closure. The cabinets also feature trilingual warning with Haz-Alert™ reflective labels, dual vents with flame arresters, adjustable leveling feet, and grounding connector.
$2,381.00 - $4,742.84CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Number of Shelves Self-Closing
Availability Pricing
Justrite 894520 Sure-Grip® EX Flammable Cabinet, Self Close, 2 Doors, 2 Shelves, 45 Gal RK-81790-36 Mfr # 894520
COMPARE
45 2 Yes
$2,534.23CAD / Each
Justrite 896020 Sure-Grip® EX Flammable Cabinet, Self Close, 2 Doors, 2 Shelves, 60 Gal RK-81790-37 Mfr # 896020
COMPARE
60 2 Yes
$3,018.98CAD / Each
Justrite 899020 Sure-Grip® EX Flammable Cabinet, Self Close, 2 Doors, 2 Shelves, 90 Gal RK-81790-38 Mfr # 899020
COMPARE
90 2 Yes
$4,720.74CAD / Each
Justrite 896000 Sure-Grip® EX Flammable Cabinet, Manual Close, 2 Doors, 2 Shelves, 60 Gal RK-81790-42 Mfr # 896000
COMPARE
60 2 No
$3,693.79CAD / Each
Justrite 899000 Sure-Grip® EX Flammable Cabinet, Manual Close, 2 Doors, 2 Shelves, 90 Gal RK-81790-43 Mfr # 899000
COMPARE
90 2 No
$4,742.84CAD / Each
Justrite 893020 Sure-Grip® EX Flammable Cabinet, Self Close, 2 Doors, 1 Shelf, 30 Gal RK-81790-45 Mfr # 893020
COMPARE
30 1 Yes
$2,381.00CAD / Each