This product is now discontinued, View Related Products
Justrite Drum Cover
Easily convert your open-top drums into fire-safe receptacles
  • Spring-loaded cover securely closes via gravity
  • TÜV certified
These red drum covers allow you to convert steel drums into safe waste receptacles. Keep facilities protected against fires from combustible trash. Features a spring-loaded, rotary latch mechanism, which allows the cover to securely close via gravity or a slight touch of the hand providing an airtight seal. This reduces the potential for the drum lid to be left open by mistake, eliminating the chance for oxygen to enter and start a fire.
Covers are manufactured of steel, with a highly chemical resistant powder-coat finish. You only need a screwdriver and wrench to attachthe cover and it can be quickly removed for emptying the drum contents.
$226.90 - $957.70CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Color Self-Closing
Availability Pricing
Justrite 26730 Drum Cover For 30 Gal. Drums RK-81792-50 Mfr # 26730
COMPARE
$226.90CAD / Each
Justrite 26750 Drum Cover for 55 Gallon Drums RK-81792-51 Mfr # 26750
COMPARE
$297.77CAD / Each
Justrite 26752 Drum Cover For 55 Gal. Drum, w/ Gasket RK-81792-52 Mfr # 26752
COMPARE
$486.22CAD / Each
Justrite 26753 Drum Cover with Vent and Gasket for 55 Gal Drum, Steel, Self-Latching RK-81792-54 Mfr # 26753
COMPARE
55 Red Yes
$957.70CAD / Each