Justrite Drip Can and Spill Tray
Contain spills while preventing potential flammable ignition
Both the spill tray and drip can are ideal for protecting contents from possible fire exposure.
The steel spill tray has a low-profile, large diameter surface to contain spills and overfill from safety cans preventing potential flammable ignition. Constructed of 24-gauge coated steel.
The drip can is ideal for placement below drum faucets using flammable liquids. Constructed of 24-gauge coated steel with a durable powder paint finish and perforated metal fire baffle, which acts as a flame arrester to protect contents from fire exposure.
$179.39 - $199.28CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Capacity (Liters)
Availability Pricing
Justrite 10177 Spill Tray, Steel, .25 Gal Capacity RK-09934-42 Mfr # 10177
COMPARE
0.5 1.89
$179.39CAD / Each
Justrite 10376 Drip Can with Handles and Fire Baffle, Steel, 1 Gal RK-09934-45 Mfr # 10376
COMPARE
1 3.79
$199.28CAD / Each