Julabo CORIO™ CD Refrigerating/Heating Circulators
Recessed cooling coil yields more usable bath space
  • Easy pump change-over from internal to external circulation
  • Securely lock in power cord for safe connection
  • Easily view bright white LED display from across the room
  • Built-in drain tap for easy and safe drainage
These CORIO™ CD refrigerating/heating circulators feature all the core functions required for daily tasks in laboratories and industry for temperature control of a wide variety of applications and samples. All operating controls and safety functions can be easily accessed from the front of unit. One power switch controls the circulator and refrigeration unit while the pump control lever, located below the bright digital LED display, makes changing between internal and external circulation simple. Inclined pump connections (M16x1) facilitate the connection of external applications.
To minimize condensation, integrated ventilation directs air over the bath lid. Specially designed pyramids on inside of the lid ensure condensed liquid drips directly back into the bath. The ergonomic plastic handle on the lid prevents direct contact with hot surfaces. Designed with a recessed cooling coil, the internal bath provides more usable space. High-temperature shutoff and low-level liquid alarm protect the bath and application from adverse events.
Constructed with no vents or connections on its sides, the circulators can be placed right next to other units, applications, or a wall, freeing up valuable lab space. Other features include a front-mounted drain tap for removing bath fluids without the use of tools, USB interface for remote control, removable front magnetic grid for easy cleaning and maintenance, lockable power plug, and integrated tank handles (front and back).
$9,104.08 - $22,229.03CAD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Power (VAC) Reservoir Capacity (L)
Availability Pricing
Julabo CORIO™ CD-200F Refrigerated/Heating Circulator, 4 L Tank, -20 to 150 °C, 1 kW Heater and 220 W at 20 °C Cooling, R134a; 115 VAC, 60 Hz RK-12149-01 Mfr # 9012701.02
COMPARE
-20 115 4
$9,104.08CAD / Each
Julabo CORIO™ CD-200F Refrigerated/Heating Circulator, 4 L Tank, -20 to 150 °C, 2 kW Heater and 220 W at 20 °C Cooling, R134a; 230 VAC, 50 Hz RK-12149-03 Mfr # 9012701.03
COMPARE
-20 230 4
$9,831.93CAD / Each
Julabo CORIO™ CD-201F Refrigerated/Heating Circulator, Low Profile, 4 L Tank, -20 to 150 °C, 1 kW Heater and 220 W at 20 °C Cooling, R134a; 115 VAC, 60 Hz RK-12149-07 Mfr # 9012702.02
COMPARE
-20 115 4
$9,104.08CAD / Each
Julabo CORIO™ CD-201F Refrigerated/Heating Circulator, Low Profile, 4 L Tank, -20 to 150 °C, 2 kW Heater and 220 W at 20 °C Cooling, R134a; 230 VAC, 50 Hz RK-12149-09 Mfr # 9012702.03
COMPARE
-20 230 4
$9,831.93CAD / Each
Julabo CORIO™ CD-300F Refrigerated/Heating Circulator, 4 L Tank, -25 to 150 °C, 1 kW Heater and 310 W at 20 °C Cooling, R134a; 115 VAC, 60 Hz RK-12149-11 Mfr # 9012703.02
COMPARE
-25 115 4
$10,573.05CAD / Each
Julabo CORIO™ CD-600F Refrigerated/Heating Circulator, 7.5 L Tank, -35 to 150 °C, 1 kW Heater and 600 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz RK-12149-17 Mfr # 9012704.02
COMPARE
-35 115 7.5
$13,313.55CAD / Each
Julabo CORIO™ CD-601F Refrigerated/Heating Circulator, 10 L Tank, -35 to 150 °C, 1 kW Heater and 600 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz RK-12149-21 Mfr # 9012705.02
COMPARE
-35 115 10
$14,265.36CAD / Each
Julabo CORIO™ CD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 1 kW Heater and 310 W at 20 °C Cooling, R290; 115 VAC, 60 Hz RK-12153-12 Mfr # 9012713.N1.02
COMPARE
-30 115 4
$10,573.05CAD / Each
Julabo CORIO™ CD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 2 kW Heater and 310 W at 20 °C Cooling, R290; 230 VAC, 60 Hz RK-12153-13 Mfr # 9012713.N1.13
COMPARE
-30 230 4
$10,573.05CAD / Each
Julabo CORIO™ CD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 2 kW Heater and 310 W at 20 °C Cooling, R290; 230 VAC, 50 Hz RK-12153-14 Mfr # 9012713.N1.03
COMPARE
-30 230 4
$11,418.77CAD / Each
Julabo CORIO™ CD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 1 kW Heater and 310 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz RK-12153-15 Mfr # 9012713.S1.02
COMPARE
-30 115 4
$10,573.05CAD / Each
Julabo CORIO™ CD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 2 kW Heater and 310 W at 20 °C Cooling, R449A; 230 VAC, 60 Hz RK-12153-16 Mfr # 9012713.S1.13
COMPARE
-30 230 4
$10,573.05CAD / Each
Julabo CORIO™ CD-310F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 2 kW Heater and 310 W at 20 °C Cooling, R449A; 230 VAC, 50 Hz RK-12153-17 Mfr # 9012713.S1.03
COMPARE
-30 230 4
$11,418.77CAD / Each
Julabo CORIO™ CD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 1 kW Heater and 450 W at 20 °C Cooling, R290; 115 VAC, 60 Hz RK-12153-18 Mfr # 9012714.N1.02
COMPARE
-30 115 4
$11,742.92CAD / Each
Julabo CORIO™ CD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 2 kW Heater and 450 W at 20 °C Cooling, R290; 230 VAC, 60 Hz RK-12153-19 Mfr # 9012714.N1.13
COMPARE
-30 230 4
$11,742.92CAD / Each
Julabo CORIO™ CD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 2 kW Heater and 450 W at 20 °C Cooling, R290; 230 VAC, 50 Hz RK-12153-20 Mfr # 9012714.N1.03
COMPARE
-30 230 4
$12,682.94CAD / Each
Julabo CORIO™ CD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 1 kW Heater and 450 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz RK-12153-21 Mfr # 9012714.S1.02
COMPARE
-30 115 4
$11,742.92CAD / Each
Julabo CORIO™ CD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 2 kW Heater and 450 W at 20 °C Cooling, R449A; 230 VAC, 60 Hz RK-12153-22 Mfr # 9012714.S1.13
COMPARE
-30 230 4
$11,742.92CAD / Each
Julabo CORIO™ CD-450F Refrigerated/Heating Circulator, 4 L Tank, -30 to 150 °C, 2 kW Heater and 450 W at 20 °C Cooling, R449A; 230 VAC, 50 Hz RK-12153-23 Mfr # 9012714.S1.03
COMPARE
-30 230 4
$12,682.94CAD / Each
Julabo CORIO™ CD-1000FW Refrigerated/Heating Circulator, Water-Cooled, 7.5 L Tank, -40 to 150 °C, 1 kW Heater and 1000 W at 20 °C Cooling, R449A; 115 VAC, 60 Hz RK-12153-24 Mfr # 9012727.02
COMPARE
-40 115 7.5
$22,229.03CAD / Each
Showing 1 - 20 of 21