This product is now discontinued, View Related Products
Jonard Precision Dynamometers
The Precision Dynamometer is a hand held force gauge for measuring small forces. Small gauges supplied with flat tip and large gauges supplied with round tip. Tolerance ±0.01 (Reading + Maximum Scale Reading)
FEATURES
  • Low cost
  • Extreme sensitivity
  • Both clockwise and counter clockwise readings
  • All gauges equipped with maximum reading pointer
$310.89 - $349.19CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (g)
Availability Pricing
Jonard Industries GD-25 Precision Dynamometer; 30-250 g RK-93949-51 Mfr # GD-25
COMPARE
250
$330.04CAD / Each
Jonard Industries GD-1 Precision Dynamometer; 0-10 g RK-93955-05 Mfr # GD-1
COMPARE
10
$349.19CAD / Each
Jonard Industries GD-10 Precision Dynamometer; 10-100 g RK-93955-06 Mfr # GD-10
COMPARE
100
$337.41CAD / Each
Jonard Industries GD-15 Precision Dynamometer; 15-150 g RK-93955-08 Mfr # GD-15
COMPARE
150
$310.89CAD / Each
Jonard Industries GD-3 Precision Dynamometer; 4-35 g RK-93955-10 Mfr # GD-3
COMPARE
35
$313.83CAD / Each
Jonard Industries GD-30 Precision Dynamometer; 40-350 g RK-93955-11 Mfr # GD-30
COMPARE
350
$337.41CAD / Each
Jonard Industries GD-5 Precision Dynamometer; 5-50 g RK-93955-13 Mfr # GD-5
COMPARE
50
$310.89CAD / Each
Showing 1- 7 of 7