Jenway® Disposable Cuvettes
No more washing
  • Eliminate cross-contamination
Single-use polystyrene cuvettes provide fast spectroscopic analysis and offer a less expensive alternative to glass and quartz cuvettes. The cuvettes are available in a wide capacity range from 70 µL to 4.5 mL and have a pathlength of 10 mm. They are suitable for analyzing the visible region and UV region except for cuvette 83070-30 which is for visible range only.
Visible wavelength % transmission: maximum from 400 to 750 nm
UV wavelength % transmission: maximum from 200 to 400 nm
$92.82 - $656.03CAD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Capacity (mL) Pathlength (mm) Description
Availability Pricing
COMPARE
4.5 10 Visible Region Disposable Plastic Cuvettes, 2.4 to 4.5 mL, 10 mm Path; 500/PK
$417.71CAD / Each
COMPARE
4.5 10 UV/Visible Region Disposable Plastic Cuvettes, 2.4 to 4.5 mL, 10 mm Path; 100/PK
$159.49CAD / Each
COMPARE
3 10 UV/Visible Region Disposable Plastic Cuvettes, 1.5 to 3 mL, 10 mm Path; 100/PK
$180.86CAD / Each
COMPARE
4.5 10 UV/Visible Region Disposable Plastic Cuvettes, 2.4 to 4.5 mL, 10 mm Path; 500/PK
$656.03CAD / Each
COMPARE
1.5 10 UV/Visible Region Disposable Plastic Cuvettes, 0.07 to 1.5 mL, 10 mm Path; 100/PK
$231.69CAD / Each
COMPARE
4.5 10 Visible Region Disposable Plastic Cuvettes, 2.4 to 4.5 mL, 10 mm Path; 100/PK
$92.82CAD / Each
COMPARE
3 10 Visbile Region Disposable Plastic Cuvette, 1.5 to 3 mL, 10 mm Path; 100/PK
$92.82CAD / Each
Showing 1- 7 of 7