Jenway® 73 Series Visible and UV/Visible Spectrophotometers
Icon-driven software and easy-to-use navigation system provide intuitive usability
  • Autologging capabilities enable repeat measurements of the same sample with a selected time interval
  • Minimal footprint with large graphical display
These Jenway® 73 series single-beam spectrophotometers have improved optics resulting in a narrow spectral bandwidth of 5 nm and absorbance range of –0.3 to 2.5 A. All models feature measurement modes for absorbance, % transmittance, and concentration. Advanced models add spectrum scanning, quantitation, and kinetics.
Visible spectrum models have a tungsten halogen lamp with a "lamp save" feature which shuts off the lamp after periods of inactivity. UV/visible models feature a “press to read” xenon lamp to provide accurate readings and extend lamp life. Connect to a PC or printer via the RS-232 interface. The included software provides easy data export and enables the spectrophotometer to be controlled by a PC.
$6,683.30 - $9,095.24CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Measurement Mode Min Wavelength (nm) Source Lamp
Availability Pricing
COMPARE
Absorbance, % transmittance, and concentration 320 Tungsten halogen
$7,191.62CAD / Each
COMPARE
Absorbance, % transmittance, and concentration 198 Xenon
$7,064.91CAD / Each
COMPARE
Absorbance, % transmittance, concentration, spectrum scanning, quantitation, and kinetics 320 Tungsten halogen
$6,683.30CAD / Each
COMPARE
Absorbance, % transmittance, concentration, spectrum scanning, quantitation, and kinetics 198 Xenon
$9,095.24CAD / Each