This product is now discontinued, View Related Products
Jenway 67 Series Graphical Visible and UV/Visible Scanning Spectrophotometers
Intuitive touch-screen LCD displays results and prompts you through programmed operations
  • Control all functions through the screen and with the touch-wheel for fine cursor control
  • Transfer data directly to your PC with included software or store and transfer data with SD/USB memory cards
  • Instrument and data access can be controlled through multi-level secure log-in
These spectrophotometers are programmed with modes for photometrics, spectrum scanning, multi-wavelength analysis, kinetics, and quantitation. Extensive post-measurement tools ensure that results are presented to your requirements.
Operational security is enabled through assigned PIN codes if desired. Individual methods can be set as personal, public (open access), or read-only for modification by the originator. Optional sample holders are available for application flexibility — the instrument recognizes each accessory compartment and automatically adjusts operation parameters.
$6,879.26 - $9,546.09CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Bandwidth Max Wavelength (nm) Min Wavelength (nm)
Availability Pricing
COMPARE
4.0 nm 1100 320
$6,879.26CAD / Each
COMPARE
4.0 nm 1100 190
$9,061.35CAD / Each
COMPARE
1.5 nm 1100 190
$9,546.09CAD / Each