International Products Corp Zymit® Low Foam Enzyme Cleaner
Ideal for use in ultrasonic cleaners and glassware washers
  • Available in 1-L bottles to 210 kg plastic drums!
This enzyme-based liquid cleaner removes biological materials from chemical, food, and pharmaceutical equipment. Cleaner has pH of 7.5 allowing you to use it with delicate materials and metals. Solution is USDA approved.
$86.34 - $852.36CAD / Case of 12
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Format Description
Availability Pricing
International Products Corp Zymit® Low-Foam Enzyme Cleaner; 12 x 1 L RK-18101-20 Mfr # Z-9701-12
COMPARE
Liquid Zymit® Low-Foam Enzyme Cleaner; 12 x 1 L
$852.36CAD / Case of 12
International Products Corp Zymit® Low-Foam Enzyme Cleaner; 1 L RK-18101-30 Mfr # Z-9701-12 1L EACH
COMPARE
Liquid Zymit® Low-Foam Enzyme Cleaner; 1 L
$86.34CAD / Each