This product is now discontinued, View Related Products
Instek GDS-300 Series Compact Portable Digital Storage Oscilloscopes
Built-in digital multimeter works simultaneously with oscilloscope functions for a complete test and measurement instrument
  • Full touch panel, 7" (18 cm) LCD allows you to easily make waveform adjustments and set trigger conditions
  • Built-in engineering calculator and trend plot enable field analysis and debugging
  • Two input channels provide versatility
  • View results in either landscape or portrait orientation
The GDS-300 series (models 20050-03 through -05) feature a 50,000-count digital multimeter for greater resolution and memory depth of 5 million points per channel for expanded logging episodes.
$2,031.80 - $2,961.51CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Bandwidth (MHz) Channels Input Impedance
Availability Pricing
GW Instek GDS-307 Portable Digital Oscilloscope with 50,000 Count DMM, 70 MHz, 2 channel RK-20050-03 Mfr # GDS-307
COMPARE
70 2 1 MΩ ±2%, 16.5 pF
$2,031.80CAD / Each
GW Instek GDS-310 Portable Digital Oscilloscope, 100 MHz, 2 channel RK-20050-04 Mfr # GDS-310
COMPARE
100 2 1 MΩ ±2%, 16.5 pF
$2,404.57CAD / Each
GW Instek GDS-320 Portable Digital Oscilloscope, 200 MHz, 2 channel RK-20050-05 Mfr # GDS-320
COMPARE
200 2 1 MΩ ±2%, 16.5 pF
$2,961.51CAD / Each