This product is now discontinued, View Related Products
Inoculating Needle and Loops
Save time and eliminate cross-contamination
  • Sterile to eliminate cross-contamination
  • Color coded for easy identification
Prevent the risk of cross-contamination with these sterile, disposable needles and loops. Packaged in boxes of 960 (32 per bag, 30 bags per box).
$146.60 - $150.29CAD / Box of 960
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Sterile Description
Availability Pricing
COMPARE
White Yes Inoculating needle and loop; microneedle; color, white; dimensions; 9\"L; 960/box
$150.29CAD / Box of 960
COMPARE
Blue Yes Inoculating Loop, Blue, 9\" Long, 1 uL/1 mm ID Loop; 960/Box
$146.60CAD / Box of 960
COMPARE
Yellow Yes Inoculating Loop, Yellow, 9\" Long, 10 uL/1 mm ID Loop; 960/Box
$150.29CAD / Box of 960