Incubator-Genie Benchtop Shaking/Rotating Incubating Shaker
Incubate, shake, rock, and rotate all in one compact unit.
  • Magnetic platform can be set to rock or rotate almost any container
  • Offers precise temperature control of ±0.2°C
  • Variety of accessories make this shaker versatile for any lab
Use this 3-in-1 unit to perform assays, washings, and hybridizations more efficiently. Air within the chamber is continuously and gently recirculated to ensure consistent and uniform temperature from 28 to 75°C. Digital control makes it simple to set rotating/rocking speed and desired temperature which are individually displayed for reproducible conditions.
A shatterproof window allows viewing of samples without disruption of chamber environment. Magnetic stainless steel platform and clip plate system hold virtually any container at any angle.
$7,864.96CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Genie SI-1401 Incubator-, Incubating Rotating Shaker; 230V RK-51603-02 Mfr # SI-1401
COMPARE
230
$7,864.96CAD / Each