This product is now discontinued, View Related Products
Incu-Shaker Incubating Shakers with MAGic Clamp Platform
Instantly change flask clamps with magnetic platform and accessories
  • Clamps utilize a powerful magnetic base to securely attach to the shaking platform without screws, bolts, or tools
  • Brushless motor is designed for years of trouble-free service, even under continuous use
These 10-L shakers offer exceptional temperature uniformity and accuracy due to a proprietary heat distribution technology that ensures temperatures are precisely maintained throughout the chamber within 0.25%, even in the corners. Digitally regulated temperature with precisely controlled orbital mixing is optimized for culturing a wide variety of cell types.
Featuring the MAGic Clamp platform, this shaker lets you customize the size and types of vessels being used by simply exchanging clamp sizes. Speed, temperature, and time are controlled with a push of a button and are displayed on the LED panel. Overtemperature and overspeed controls ensure safety, long life, and sample integrity. Advanced motor isolation system minimizes noise and vibration.
Available in heated or heated/refrigerated models, the 10-L incubating shaker easily handles your heavy workloads in the lab. A large 18" x 18" (45.7 x 45.7 cm) platform is perfect for high-capacity applications.
$9,127.65 - $16,008.38CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
Benchmark Scientific H1010-CP Incu-Shaker 10L with Universal MAGic Clamp Platform, 115 VAC. RK-51705-11 Mfr # H1010-CP
COMPARE
115 Incu-Shaker 10L with Universal MAGic Clamp Platform, 115 VAC.
$11,860.79CAD / Each
Benchmark Scientific H1010-E-CP Incu-Shaker 10L with Universal MAGic Clamp Platform, 230 VAC. RK-51705-12 Mfr # H1010-E-CP
COMPARE
230 Incu-Shaker 10L with Universal MAGic Clamp Platform, 230 VAC.
$9,127.65CAD / Each
Benchmark Scientific H1012-CP Incu-Shaker 10L Refrigerated with Universal MAGic Clamp Platform, 115 VAC. RK-51705-13 Mfr # H1012-CP
COMPARE
115 Incu-Shaker 10L Refrigerated with Universal MAGic Clamp Platform, 115 VAC.
$16,008.38CAD / Each
Benchmark Scientific H1012-E-CP Incu-Shaker 10L Refrigerated with Universal MAGic Clamp Platform, 230 VAC. RK-51705-14 Mfr # H1012-E-CP
COMPARE
230 Incu-Shaker 10L Refrigerated with Universal MAGic Clamp Platform, 230 VAC.
$16,008.38CAD / Each