In-Line Flow Indicators for Liquids and Gases

An inexpensive way to visually monitor flow of liquids and gases
Indicators give visual confirmation of flow through a clear polystyrene housing. All models can be used with noncorrosive liquids and gases. Connect tubing to indicators via hose barb connections; model 06297-05 has straight ends. All models are pressure rated to 10 psig.
$35.66 - $74.41CAD / Pkg of 1

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Pipe Size
Availability Pricing
Dynalon Ball Flow Indicator, 0.2 to 6.5 L/min for 1/4 to 3/8" ID Tubing RK-06297-00 Mfr # 227155
COMPARE
1/4 to 3/8" ID Tubing
$74.41CAD / Pkg of 1
Bel-Art 19937-0001 In-Line Flow Indicators, 0.5 to 10 L/min; Paddle Flow, 3/16" to 1/4" Tubing RK-06297-05 Mfr # 19937-0001
COMPARE
3/16" to 1/4" ID Tubing
$35.66CAD / Pkg of 1
Bel-Art 19937-0002 Paddle Flow Indicator, 0.5 to 10 L/min for 1/4" to 5/16" ID Tubing RK-06297-06 Mfr # 19937-0002
COMPARE
1/4" to 5/16" ID Tubing
$35.66CAD / Pkg of 1
Bel-Art 19937-0003 Paddle Flow Indicator, 0.5 to 10 L/min for 5/16 to 1/2" ID Tubing RK-06297-07 Mfr # 19937-0003
COMPARE
5/16 to 1/2" ID Tubing
$35.66CAD / Pkg of 1
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat