This product is now discontinued, View Related Products
In-Line Carbon Filter Capsules
Disposable filter capsule contains high-purity activated carbon and glass microfiber HEPA filter
  • Large surface area of activated carbon for maximum flow and contaminant removal
These in-line filters are ideal for applications involving compressed air lines, vacuum pumps, and instrument exhaust emissions. The capsule contains a high-purity granular activated carbon and has a pleated glass microfiber HEPA filter at the base of the unit which retains 99.97% of all particles greater than 0.3 µm. Device features a high-purity polypropylene housing.
Typical applications include water, chemical and reagent purification, odor control, vacuum pump outlets, decolorizing, and oil clarification. Maximum operating pressure is 60 psi (4.1 bar).
$295.41 - $322.08CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connections Filter Size
Availability Pricing
ZenPure In-line Carbon Filter Capsule, 96 x 66 mm RK-49708-10 Mfr # SKCHGHS4H4H
COMPARE
1/2\" hose barb inlet 3/8\" hose barb outlet 155 x 70
$295.41CAD / Each
ZenPure In-line Carbon Filter Capsule, 183 x 66 mm RK-49708-12 Mfr # SKCHGSL4H4H
COMPARE
1/2\" hose barb inlet 3/8\" hose barb outlet 210 x 70
$322.08CAD / Each