This product is now discontinued, View Related Products
Imada ZTS Series Digital Force Gauges
Expanded push/pull capacity of 1100 lbs provides additional flexibility
  • Electro-luminescent display is highly visible in dark locations
  • Processes and transmits 2000 data points per second
  • Color-coded LED indicators and output signal for Go/No-Go testing
  • Ergonomic design with nonslip grips, and heavy-duty metal body construction
  • 1000 data records of memory for recall or download
  • External switch display, hold and clear
  • User-selectable real time and peak mode
  • Overload alarm and output signal
  • Multilanguage program menus
  • Low-battery indicator
These digital force gauges allow you to easily configure the display to show the basic force value, unit, and peak, or display in multiline format which has two sections of user-selectable data. Select units, reverse the display, high/low set points, or a variety of other functions from the intuitive screen menu. The gauges incorporate LED indicators which alert you to Go/No-Go conditions (orange = less than low set point; green = between high and low; red = over the high set point).
The ZT Logger data acquisition software downloads data from the gauge memory or captures a continuous stream of data at 10 data points per second. Additional software features include gauge reset, Min/Max, avg, standard deviation, count values over the high set point, save/add data to .csv files, and gauge program/save capability.
$1,510.22 - $2,048.01CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Capacity
Availability Pricing
Imada ZTS-0.4 Gauge, 7 oz., 200 g., 2 N RK-25302-81 Mfr # ZTS-0.4
COMPARE
7 oz
$1,510.22CAD / Each
Imada ZTS-1 Gauge, 18 oz., 500 kg., 5 N RK-25302-82 Mfr # ZTS-1
COMPARE
18 oz
$1,601.57CAD / Each
Imada ZTS-4 Gauge, 4.4 lb., 2 kg., 20 N RK-25302-83 Mfr # ZTS-4
COMPARE
4.4 lbs
$1,510.22CAD / Each
Imada ZTS-11 Gauge, 11 lb., 5 g., 50 N RK-25302-84 Mfr # ZTS-11
COMPARE
11 lbs
$1,510.22CAD / Each
Imada ZTS-44 Gauge, 44 lb., 20 kg., 200 N RK-25302-86 Mfr # ZTS-44
COMPARE
44 lbs
$1,601.57CAD / Each
Imada ZTS-110 Gauge, 110 lb., 50 kg., 500 N RK-25302-87 Mfr # ZTS-110
COMPARE
110 lbs
$1,598.63CAD / Each
Imada ZTS-220 Gauge, 220 lb., 100 kg., 1000 N RK-25302-88 Mfr # ZTS-220
COMPARE
220 lbs
$1,672.30CAD / Each
Imada ZTS-550 Gauge, 440 lb., 200 kg., 2500 N RK-25302-89 Mfr # ZTS-550
COMPARE
440 lbs
$1,930.14CAD / Each
Imada ZTS-1100 Gauge, 1100 lb., 500 kg., 5000 N RK-25302-90 Mfr # ZTS-1100
COMPARE
1100 lbs
$2,048.01CAD / Each
Showing 1- 9 of 9