This product is now discontinued, View Related Products
IKA ULTRA TURRAX Tube Disperser 47981
Closed homogenizing system – homogenize samples inside disposable sample tubes
  • Hygienic and clean homogenizing in closed tube system
  • High level of user safety
  • Process sample volumes from 2 - 50 mL
This unique compact homogenizer processes samples inside disposable hermetically sealed tubes, which prevents sample contamination. Each sample tube contains a stirring device, blade, or ball for mixing and homogenizing small samples. Disposable tubes lock into place and remove quickly with a simple twist. Choose from three tube styles with different mixing devices to process your particular sample type.
$147.34 - $2,024.44CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm)
Availability Pricing
IKA 3827500 ULTRA TURRAX Tube Disperser, Control version, 100-240 VAC, 50/60 Hz RK-04738-87 Mfr # 3827500
COMPARE
6000
$2,024.44CAD / Each
IKA 3645000 ULTRA TURRAX Tube Disperser, basic, 100-240 VAC, 50/60 Hz RK-04738-89 Mfr # 3645000
COMPARE
6000
$1,644.30CAD / Each
IKA 3553501 Ultra Turrax Tube Drive, Yellow Line 115 Vac RK-04738-91 Mfr # 3553501
COMPARE
$844.25CAD / Each
IKA 3553500 Ultra Turrax Tube Drive, Yellow Line 230 Vac RK-04738-92 Mfr # 3553500
COMPARE
$844.25CAD / Each
IKA ULTRA TURRAX Ball-Mill Tube, Ceramic, BMT-20 K; 10/pack RK-04739-00 Mfr # 3599700-10
COMPARE
$147.34CAD / Pkg