This product is now discontinued, View Related Products
IKA RW 47 Digital Pilot-Process Mixer
Powerful mechanically-controlled mixer for highly viscous applications in labs, pilot plants, and small-scale production
  • Two speed ranges for intensive mixing up to 100,000 cps
  • Stirs volumes up to 52 gallons (200 liters)
These mechanically controlled mixers are designed for intensive mixing applications in laboratories and pilot plants. Easy-to-use controls and a clear LED display of rpm enable you to precisely control and repeat your process parameters. Mixer features infinitely adjustable dual-speed control.
Self-locking temperature limiter protects against overheating and ensures safe operation through thermal overload shut-off. Quiet operation and a robust, ergonomic design ensure a long service life.
$16,612.47 - $28,275.83CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
IKA 4050010 Digital Mixer, 230V, 60 Hz, 3 phase RK-50702-80 Mfr # 4050010
COMPARE
1300 4248 230 VAC, 3-Phase
$16,612.47CAD / Each
IKA 0010000663 Digital Mixer Package; 230V, 60 Hz, 3 phase RK-50702-85 Mfr # 0010000663
COMPARE
1300 4248 230 VAC, 3-Phase
$28,275.83CAD / Each