IKA Rocker Shakers
Three-dimensional tumbling motion for smooth mixing tasks
These rocking shakers provide homogeneous mixing in flasks, Petri dishes and tubes. Ideal for the cultivation of cells, DANN extractions, distributing of cells, or staining and de-staining of cells. Choose either the basic or digital shaker; both have simple keypad control, anti-slip rubber mat, continuous operation, and constant shaking speed even with load and voltage fluctuations. The digital model offers the addition of an LED display, adjustable speeds, programmable timer, and adjustable rocking angle.
$2,329.43CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm)
Availability Pricing
IKA 4001000 Rocker Shaker, Digital (11"W x 6 1/2"H x x13"D) RK-04304-15 Mfr # 0004001000
COMPARE
80
$2,329.43CAD / Each