IKA EUROSTAR 200 Control P4 Digital Mixers with Wireless Remote
Wireless controller lets you control and monitor all functions remotely
  • EUROSTAR 200 Control P4 Dual-Range/High-Torque Mixer will mix volumes up to 26.4 gallons (100 liters) and viscosities up to 100,000 cps
  • A high-torque rating and two selectable speed ranges give you the option of high viscosity or high-speed mixing
  • Ultra-clear, high-contrast LCD controller screen displays all process parameters
  • Torque trend function measures viscosity changes
  • Integrated temperature measurement lets you monitor fluid temperature
These powerful and advanced mixers handle volumes up to 26.4 gallons (100 liters) and viscosities up to 100,000 cps. Unique features include a removable wireless controller with a multilingual LCD that lets you control and monitor all mixer functions. An integrated torque trend display is provided for the measurement of viscosity changes and a 1000 Pt RTD temperature sensor input lets you monitor and display fluid temperature. Mixers also include RS-232 and USB interfaces to control and document all parameters.
Additional control features include a timer function that allows you to preset run times and an interval function that enables you to program pause/run cycles.
Mixers feature infinitely adjustable stepless speed control. Continuous comparison of shaft speed to desired speed is maintained and variations are adjusted automatically. This guarantees a constant speed even with changes in fluid viscosity.
Quiet, fan-cooled motors feature thermal overload shut-off, and an efficient heat-dissipating design to ensure a long service life. Motor is maintenance-free and rated for continuous duty. Safety circuits ensure automatic cut-off in an anti-stall or overload condition.
$9,758.26CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Max Volume (Liters)
Availability Pricing
IKA 4090001 Digital P4 Mixer, High-Torque Dual-Range, 115VAC RK-50705-60 Mfr # 4090001
COMPARE
530 935 100
$9,758.26CAD / Each