This product is now discontinued, View Related Products
IDEX VacuTight™ Chromatography Fittings with M6 Flat-Bottom Connection
Make dependable connections under vacuum and low pressure conditions
  • Small size ensures sufficient nut/thread engagement
  • Design can allow a large number of connections in a small space
  • Vacuum applications rated up to 25" Hg (84 kPa)
VacuTight fittings are ideal for clinical diagnostic, high throughput screening, and other automated liquid-handling applications due to their small size and dependable, airtight connections in low pressure and vacuum environments. The flat-bottom ferrule configuration impedes tubing inner diameter reduction and overcompression that can be a problem with many coned fittings.
$21.22 - $262.26CAD / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex P-931X VacuTight™ Fitting, Standard, Red Acetal, 1/16" OD Tubing, M6 Flat Bottom; 10/PK RK-02017-56 Mfr # P-931X
COMPARE
1/16 Delrin® acetal VacuTight™ Fitting, Standard, Red Acetal, 1/16\" OD Tubing, M6 Flat Bottom; 10/PK
$232.80CAD / Pkg of 10
Idex P-933X VacuTight™ Fitting, Short, Red Acetal, 1/16" OD Tubing, M6 Flat Bottom; 10/PK RK-02017-58 Mfr # P-933X
COMPARE
1/16 Delrin® acetal VacuTight™ Fitting, Short, Red Acetal, 1/16\" OD Tubing, M6 Flat Bottom; 10/PK
$226.90CAD / Pkg of 10
Idex P-945 VacuTight™ Fitting, Short, Red Acetal, 1/8" OD Tubing, M6 Flat Bottom; 1/EA RK-02017-69 Mfr # P-945
COMPARE
1/8 Delrin® acetal VacuTight™ Fitting, Short, Red Acetal, 1/8\" OD Tubing, M6 Flat Bottom; 25/PK
$21.22CAD / Each
Idex P-945X VacuTight™ Fitting, Short, Red Acetal, 1/8" OD Tubing, M6 Flat Bottom; 10/PK RK-02017-71 Mfr # P-945X
COMPARE
1/8 Delrin® acetal VacuTight™ Fitting, Short, Red Acetal, 1/8\" OD Tubing, M6 Flat Bottom; 10/PK
$262.26CAD / Pkg of 10
Idex P-947X VacuTight™ Fitting, Headless, Red Acetal, 1/8" OD Tubing, M6 Flat Bottom; 10/PK RK-02017-73 Mfr # P-947X
COMPARE
1/8 Delrin® acetal VacuTight™ Fitting, Headless, Red Acetal, 1/8\" OD Tubing, M6 Flat Bottom; 10/PK
$226.90CAD / Pkg of 10