Hydrogen (H2) Gas Mass Flow Controllers with Metal bodies
Precisely control flow rates with built-in valve
  • Proven design uses metal body components for added durability
  • Accumulated flow can be logged using an optional totalizer
  • Includes NIST-traceable certificate
These controllers feature an advanced U-shaped sensor that ensures accurate and repeatable results. Gas flow measurements are unaffected by moderate temperature and pressure variations at the inlet. The meter also allows a four-point calibration across the flow range to improve meter linearity.
The flow rate set point can be established by either a local potentiometer or by a remote 4 to 20 mA or 0 to 5 VDC signal. Output data is sent via a 0 to 5 VDC or 4 to 20 mA signal; an analog-to-RS converter is also available for data collection and analysis on your PC.
All controllers include a detachable 3-1/2-digit LCD that can be tilted up to 90° for easy reading. The display is remote mountable to three feet using extension cable 32662-70. The totalizer option 32650-22 will display total accumulated flow.
The controller is protected from polarity reversal or short circuit by a built-in resettable fuse. Aluminum models have wetted materials of anodized aluminum, brass, and Viton; stainless steel (SS) models have wetted materials of SS and Viton.
Response Time:
Generally 2 seconds to within 2% of actual flow rate over 25 to 100% of full scale.
Accuracy:
These precalibrated flow controllers operate at inlet pressures between 5 and 60 psi and at gas temperatures between 59 and 77°F (15 to 25°C) while maintaining the stated percent full-scale accuracy and linearity. When operating beyond 5 to 60 psi, add ±0.01%/psi full-scale; if operating beyond 59 to 77°F (15 to 25°C), add ±0.15%/°C full-scale.
$1,386.46 - $1,900.67CAD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (LPM) Process Connection Wetted Materials
Availability Pricing
Aalborg GFCS-011162 Compact Gas Mass Flow Controller, 0-10 sccm, H2, Aluminum Body RK-32660-52 Mfr # GFCS-011162
COMPARE
0.01 1/4" Compression Anodized Aluminum, Brass, Viton
$1,386.46CAD / Each
Aalborg GFCS-010030 Compact Gas Mass Flow Controller, 0-50 sccm, H2, Aluminum Body RK-32660-56 Mfr # GFCS-010030
COMPARE
0.05 1/4" Compression Anodized Aluminum, Brass, Viton
$1,386.46CAD / Each
Aalborg GFCS-010031 Compact Gas Mass Flow Controller, 0-100 sccm, H2, Aluminum Body RK-32660-58 Mfr # GFCS-010031
COMPARE
0.1 1/4" Compression Anodized Aluminum, Brass, Viton
$1,386.46CAD / Each
Aalborg GFCS-010853 Compact Gas Mass Flow Controller, 0-200 sccm, H2, Aluminum Body RK-32660-60 Mfr # GFCS-010853
COMPARE
0.2 1/4" Compression Anodized Aluminum, Brass, Viton
$1,386.46CAD / Each
Aalborg GFCS-010033 Compact Gas Mass Flow Controller, 0-500 sccm, H2, Aluminum Body RK-32660-62 Mfr # GFCS-010033
COMPARE
0.5 1/4" Compression Anodized Aluminum, Brass, Viton
$1,386.46CAD / Each
Aalborg GFCS-010034 Compact Gas Mass Flow Controller, 0-1 LPM, H2, Aluminum Body RK-32660-64 Mfr # GFCS-010034
COMPARE
1 1/4" Compression Anodized Aluminum, Brass, Viton
$1,386.46CAD / Each
Aalborg GFCS-010035 Compact Gas Mass Flow Controller, 0-2 LPM, H2, Aluminum Body RK-32660-66 Mfr # GFCS-010035
COMPARE
2 1/4" Compression Anodized Aluminum, Brass, Viton
$1,386.46CAD / Each
Aalborg GFCS-010036 Compact Gas Mass Flow Controller, 0-5 LPM, H2, Aluminum Body RK-32660-68 Mfr # GFCS-010036
COMPARE
5 1/4" Compression Anodized Aluminum, Brass, Viton
$1,386.46CAD / Each
Aalborg GFCS-010037 Compact Gas Mass Flow Controller, 0-15 LPM, H2, Aluminum Body RK-32660-71 Mfr # GFCS-010037
COMPARE
15 1/4" Compression Anodized Aluminum, Brass, Viton
$1,645.78CAD / Each
Aalborg GFCS-012387 Compact Gas Mass Flow Controller, 0-30 LPM, H2, Aluminum Body RK-32660-72 Mfr # GFCS-012387
COMPARE
30 1/4" Compression Anodized Aluminum, Brass, Viton
$1,645.78CAD / Each
Aalborg GFCS-010039 Compact Gas Mass Flow Controller, 0-100 LPM, H2, Aluminum Body RK-32660-76 Mfr # GFCS-010039
COMPARE
100 3/8" Compression Anodized Aluminum, Brass, Viton
$1,900.67CAD / Each
Aalborg GFCS-010080 Compact Gas Mass Flow Controller, 0-10 sccm, H2, Stainless Steel Body RK-32661-52 Mfr # GFCS-010080
COMPARE
0.01 1/4" Compression Stainless Steel, Viton
$1,601.57CAD / Each
Aalborg GFCS-010082 Compact Gas Mass Flow Controller, 0-50 sccm, H2, Stainless Steel Body RK-32661-56 Mfr # GFCS-010082
COMPARE
0.05 1/4" Compression Stainless Steel, Viton
$1,601.57CAD / Each
Aalborg GFCS-010083 Compact Gas Mass Flow Controller, 0-100 sccm, H2, Stainless Steel Body RK-32661-58 Mfr # GFCS-010083
COMPARE
0.1 1/4" Compression Stainless Steel, Viton
$1,601.57CAD / Each
Aalborg GFCS-010084 Compact Gas Mass Flow Controller, 0-200 sccm, H2, Stainless Steel Body RK-32661-60 Mfr # GFCS-010084
COMPARE
0.2 1/4" Compression Stainless Steel, Viton
$1,601.57CAD / Each
Aalborg GFCS-010085 Compact Gas Mass Flow Controller, 0-500 sccm, H2, Stainless Steel Body RK-32661-62 Mfr # GFCS-010085
COMPARE
0.5 1/4" Compression Stainless Steel, Viton
$1,601.57CAD / Each
Aalborg GFCS-010086 Compact Gas Mass Flow Controller, 0-1 LPM, H2, Stainless Steel Body RK-32661-64 Mfr # GFCS-010086
COMPARE
1 1/4" Compression Stainless Steel, Viton
$1,601.57CAD / Each
Aalborg GFCS-010087 Compact Gas Mass Flow Controller, 0-2 LPM, H2, Stainless Steel Body RK-32661-66 Mfr # GFCS-010087
COMPARE
2 1/4" Compression Stainless Steel, Viton
$1,601.57CAD / Each
Showing 1- 18 of 18